Dihexyl sulfide ≥98.0%

Supplier: TCI America
Warning

Synonyms: Di-n-hexylsulfide, Di-n-hexylsulphide

H0377-250G H0377-25G
TCH0377-250GEA 638.32 CAD
TCH0377-250G TCH0377-025G
Dihexyl sulfide ≥98.0%
Dihexyl sulphide

CAS Number: 6294-31-1
MDL Number: MFCD00009528
Molecular Formula: C12H26S
Molecular Weight: 202.40
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Flash Point (°C): 116
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Dihexyl Sulfide

Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 260 °C
Melting Pt: –28.3 °C
Density: 0.85 (20 °C)
Flash Pt: 116 °C
MDL Number: MFCD00009528
CAS Number: 6294-31-1
EINECS: 228-556-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00151a1512889fbc0ce2678cac17cc2887cc5a13&vid=00151a1512889fbc0ce2678cac17cc2887cc5a13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR