(3-Hydroxyphenyl)acetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H0356-25G H0356-5G
TCH0356-025GEA 257.54 CAD
TCH0356-025G TCH0356-005G
(3-Hydroxyphenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
(3-Hydroxyphenyl)acetic acid

CAS Number: 621-37-4
MDL Number: MFCD00004337
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130

Formula: (C₆H₄(OH))CH₂COOH
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 130 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004337
CAS Number: 621-37-4
EINECS: 210-684-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7921d9d64c1cdaeca300ff8391f10b7376dc8077&vid=7921d9d64c1cdaeca300ff8391f10b7376dc8077&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR