Dodecyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
H0352-25G
H0352-500G
TCH0352-025GEA
150.1
CAD
TCH0352-025G
TCH0352-500G
Dodecyl-4-hydroxybenzoate ≥98.0% (by HPLC, titration analysis)
Dodecyl 4-hydroxybenzoate
CAS Number: 2664-60-0
MDL Number: MFCD00016473
Molecular Formula: C19H30O3
Molecular Weight: 306.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 55
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Dodecyl Ester, 4-Hydroxybenzoic Acid Lauryl Ester, Lauryl 4-Hydroxybenzoate, Laurylparaben
Formula:
C₁₉H₃₀O₃ Melting Pt: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016473 CAS Number: 2664-60-0 EINECS: 220-195-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...