trans-2-Hexen-1-ol ≥95.0%

Supplier: TCI America

Synonyms: trans-Hex-2-en-1-ol

H0346-25G H0346-250G
TCH0346-025GEA 54.64 CAD
TCH0346-025G TCH0346-250G
trans-2-Hexen-1-ol ≥95.0%
trans-2-Hexen-1-ol

CAS Number: 928-95-0
MDL Number: MFCD00002927
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 61
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: trans-2-Hexen-1-yl Alcohol, trans-1-Hydroxy-2-hexene

Formula: C₆H₁₂O
Boiling Pt: 162 °C
Melting Pt: 65…67 °C
Density: 0.85 (20 °C)
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002927
CAS Number: 928-95-0
EINECS: 213-191-2
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45a9f1d2409e9d6c4aa16a488a326a4a1d809f68&vid=45a9f1d2409e9d6c4aa16a488a326a4a1d809f68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR