(2-Hydroxyphenyl)acetic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Hydroxyphenylacetic acid

H0340-5G H0340-25G
TCH0340-005GEA 26.86 CAD
TCH0340-005G TCH0340-025G
(2-Hydroxyphenyl)acetic acid ≥98.0%
(2-Hydroxyphenyl)acetic acid

CAS Number: 614-75-5
MDL Number: MFCD00004323
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 147

Formula: (C₆H₄OH)CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 328
Melting Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004323
CAS Number: 614-75-5
EINECS: 210-393-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa3221b6c4e160d2fbb369eb6ece55ce807d6f58&vid=fa3221b6c4e160d2fbb369eb6ece55ce807d6f58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR