Homovanillic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

H0339-5G H0339-1G H0339-25G
TCH0339-005GEA 247.1 CAD
TCH0339-005G TCH0339-1G TCH0339-25G
Homovanillic acid ≥98.0% (by titrimetric analysis)
Homovanillic acid

CAS Number: 306-08-1
MDL Number: MFCD00004350
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 143


Other Product Names / Synonyms: Homovanillic Acid

Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 143 °C
MDL Number: MFCD00004350
CAS Number: 306-08-1
EINECS: 206-176-7
Merck Index: 13,04761

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR