3-Pyridone ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol

H0331-25G H0331-500G H0331-100G
TCH0331-025GEA 74.26 CAD
TCH0331-025G TCH0331-500G TCH0331-100G
3-Pyridone ≥98.0%
3-Pyridone

CAS Number: 109-00-2
MDL Number: MFCD00006378
Molecular Formula: C5H5NO
Molecular Weight: 95.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 126
Flash Point (°C): 173


Other Product Names / Synonyms: 3-Pyridinol, 3-Pyridone

Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 319
Melting Pt: 126 °C
Density: 1.2 g/cm³ (–70 °C)
Flash Pt: 173 °C
Storage Temperature: Ambient
MDL Number: MFCD00006378
CAS Number: 109-00-2
EINECS: 203-637-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f580dbcc4b6565ccf49a62f55bde9b2d6651f673&vid=f580dbcc4b6565ccf49a62f55bde9b2d6651f673&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR