Hypoxanthine ≥98.0% (by HPLC and UV method)
Supplier: TCI America
H0311-25G
H0311-5G
TCH0311-025GEA
112.18
CAD
TCH0311-025G
TCH0311-5G
Hypoxanthine ≥98.0% (by HPLC and UV method)
Hypoxanthine
CAS Number: 68-94-0
MDL Number: MFCD00005725
Molecular Formula: C5H4N4O
Molecular Weight: 136.11
Purity/Analysis Method: >98.0% (HPLC,E)
Form: Crystal
Formula:
C₅H₄N₄O Melting Pt: > 310 °C |
MDL Number:
MFCD00005725 CAS Number: 68-94-0 EINECS: 200-697-3 Merck Index: 13,04895 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...