2-Quinolinol ≥98.0%
Supplier: TCI America
|
Warning
|
H0304-1G
H0304-10G
TCH0304-001GEA
31.6
CAD
TCH0304-001G
TCH0304-010G
2-Quinolinol ≥98.0%
2-Quinolinol
CAS Number: 59-31-4
MDL Number: MFCD00006743
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 197
Other Product Names / Synonyms: Carbostyril, 2-Hydroxyquinoline
Formula:
C₉H₇NO Boiling Pt: 347 °C (1013 hPa) Melting Pt: 197 °C Density: 1.37 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006743 CAS Number: 59-31-4 EINECS: 200-420-6 Merck Index: 13,01835 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...