trans-4-Hydroxy-L(-)-proline ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH

H0296-25G H0296-5G H0296-100G
TCH0296-025GEA 119.08 CAD
TCH0296-025G TCH0296-5G TCH0296-100G
trans-4-Hydroxy-L(-)-proline ≥99.0% (by HPLC, titration analysis)
trans-4-Hydroxy-L(-)-proline

CAS Number: 51-35-4
MDL Number: MFCD00064320
Molecular Formula: C5H9NO3
Molecular Weight: 131.13
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -75.5 deg (C=4, H2O)


Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: L-Hydroxyproline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic Acid, H-Hyp-OH

Formula: C₅H₉NO₃
MW: 131.13 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00064320
CAS Number: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6641c01b551f7d25e459a1ed979f5da62b0833bf&vid=6641c01b551f7d25e459a1ed979f5da62b0833bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR