(4-Hydroxyphenyl)acetic acid 99,0

Supplier: TCI America
Warning

Synonyms: 4-Hydroxyphenylacetic Acid

H0290-500G H0290-25G H0290-100G
TCH0290-500GEA 322.32 CAD
TCH0290-500G TCH0290-025G TCH0290-100G
(4-Hydroxyphenyl)acetic acid 99,0
(4-Hydroxyphenyl)acetic acid

CAS Number: 156-38-7
MDL Number: MFCD00004347
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 150

Formula: (C₆H₄(OH))CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 347
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004347
CAS Number: 156-38-7
EINECS: 205-851-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff306a7d22d97fbae3f35eea6bca0c376dfa515e&vid=ff306a7d22d97fbae3f35eea6bca0c376dfa515e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR