3-Hydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Hydroxy-naphthalene-2-carboxylic acid, 2-Hydroxy-3-naphthoic acid

H0281-500G H0281-25G
TCH0281-500GEA 66.36 CAD
TCH0281-500G TCH0281-025G
3-Hydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-2-naphthoic acid

CAS Number: 92-70-6
MDL Number: MFCD00004103
Molecular Formula: C11H8O3
Molecular Weight: 188.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 220
Flash Point (°C): 230
Lambda max.: 395 nm (H2O (pH 3.5))


Other Product Names / Synonyms: 3-Hydroxy-2-naphthalenecarboxylic Acid

Formula: C₁₁H₈O₃
MW: 188.18 g/mol
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 220 °C
Flash Pt: 230 °C
MDL Number: MFCD00004103
CAS Number: 92-70-6
EINECS: 202-180-8
Merck Index: 13,04860
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dae201406d6dec229ea77e336170c4499c70efd2&vid=dae201406d6dec229ea77e336170c4499c70efd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR