1-Hydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H0279-25G H0279-500G
TCH0279-025GEA 52.93 CAD
TCH0279-025G TCH0279-500G
1-Hydroxy-2-naphthoic acid ≥98.0% (by HPLC, titration analysis)
1-Hydroxy-2-naphthoic acid

CAS Number: 86-48-6
MDL Number: MFCD00003960
Molecular Formula: C11H8O3
Molecular Weight: 188.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 208
Flash Point (°C): 150


Other Product Names / Synonyms: 1-Hydroxy-2-naphthalenecarboxylic Acid

Formula: C₁₁H₈O₃
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 208 °C
Flash Pt: 150 °C
MDL Number: MFCD00003960
CAS Number: 86-48-6
EINECS: 201-674-0
UN: 2811
ADR: 6.1,III
Merck Index: 13,04859
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=354e4b542046fe5180dab5ef688f2f1604d3834e&vid=354e4b542046fe5180dab5ef688f2f1604d3834e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR