2-Hydroxy-1-naphthaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0275-25G H0275-100G
TCH0275-025GEA 148.52 CAD
TCH0275-025G TCH0275-100G
2-Hydroxy-1-naphthaldehyde ≥98.0% (by GC, titration analysis)
2-Hydroxy-1-naphthaldehyde

CAS Number: 708-06-5
MDL Number: MFCD00004005
Molecular Formula: C11H8O2
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 192
Melting point (°C): 81


Other Product Names / Synonyms: 2-Naphthol-1-aldehyde

Formula: C₁₁H₈O₂
Boiling Pt: 192 °C
Melting Pt: 81 °C
MDL Number: MFCD00004005
CAS Number: 708-06-5
EINECS: 211-902-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f821cbfb4d0be07ca56d8b176a4c6a507faf9f7&vid=0f821cbfb4d0be07ca56d8b176a4c6a507faf9f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR