5-(Hydroxymethyl)-2-furaldehyde ≥95.0% stabilized
Supplier: TCI America
|
Warning
|
Stabilization:
Stabilised with Stabilised
H0269-1G
H0269-5G
TCH0269-001GEA
28.91
CAD
TCH0269-001G
TCH0269-005G
5-(Hydroxymethyl)-2-furaldehyde ≥95.0% stabilized
5-(Hydroxymethyl)-2-furaldehyde
(stabilized with Water)
CAS Number: 67-47-0
MDL Number: MFCD00003234
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 79
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 5-(Hydroxymethyl)furfural
Formula:
C₆H₆O₃ Boiling Pt: 292 °C (1013 hPa) Melting Pt: 27…30 °C Density: 1.24 g/cm³ (20 °C) Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003234 CAS Number: 67-47-0 EINECS: 200-654-9 Merck Index: 13,04857 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...