(±)-4-Hydroxy-3-methoxymandelic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Hydroxy-3-methoxy-DL-mandelic acid, VMA, Vanillyl mandelic acid, DL-4-Hydroxy-3-methoxymandelic acid, (±)-4-Hydroxy-3-methoxyphenylglycolic acid, 2-Hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid, Vanillylmandelic acid

H0268-1G H0268-5G
TCH0268-001GEA 72.85 CAD
TCH0268-001G TCH0268-5G
(±)-4-Hydroxy-3-methoxymandelic acid ≥98.0%
(±)-4-Hydroxy-3-methoxymandelic acid

CAS Number: 55-10-7
MDL Number: MFCD00004235
Molecular Formula: C9H10O5
Molecular Weight: 198.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 133


Other Product Names / Synonyms: Vanillylmandelic Acid

Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Boiling Pt: 421
Melting Pt: 133 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004235
CAS Number: 55-10-7
EINECS: 200-224-0
Merck Index: 12,10070
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0527df320b31b302ce2f757ac839f7c7840f87aa&vid=0527df320b31b302ce2f757ac839f7c7840f87aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR