trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid
CAS Number: 537-98-4
MDL Number: MFCD00004400
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Antioxidants, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: trans-4-Hydroxy-3-methoxycinnamic Acid
Formula:
C₁₀H₁₀O₄ MW: 194.18 g/mol Melting Pt: 173 °C |
MDL Number:
MFCD00004400 CAS Number: 537-98-4 EINECS: 208-679-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...