3-Hydroxy-p-anisaldehyde ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

H0263-250G H0263-25G
TCH0263-250GEA 398.16 CAD
TCH0263-250G TCH0263-025G
3-Hydroxy-p-anisaldehyde ≥98.0% (by titrimetric analysis)
3-Hydroxy-p-anisaldehyde

CAS Number: 621-59-0
MDL Number: MFCD00003369
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 114
Flash Point (°C): 110


Other Product Names / Synonyms: 3-Hydroxy-4-methoxybenzaldehyde, iso-Vanillin

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 114 °C
Density: 1.401 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8720dc4a42c4e88cdba6c98722c6bc411b22dda&vid=9f5f5629e329bc5bb950624cb2aa34e70a90ce40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR