4'-Hydroxy-3'-methoxyacetophenone ≥98.0%
Supplier: TCI America
Synonyms:
4-Hydroxy-3-methoxy-acetophenone, 1-(4-Hydroxy-3-methoxyphenyl)ethanone, Acetovanillone, Apocynin
CAS Number: 498-02-2
MDL Number: MFCD00008747
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 115
Flash Point (°C): 113
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, ROS/RNS-related Enzyme Inhibitors, Others (Phytochemicals)
Other Product Names / Synonyms: Acetovanillone, Apocynin
Formula:
C₉H₁₀O₃ Boiling Pt: 300 °C Melting Pt: 115 °C Flash Pt: 113 °C |
MDL Number:
MFCD00008747 CAS Number: 498-02-2 EINECS: 207-854-5 Merck Index: 13,00746 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...