5-Hydroxyindole ≥98.0%
Supplier: TCI America
Synonyms:
1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indole
H0252-5G
H0252-1G
H0252-100MG
TCH0252-5GEA
271.76
CAD
TCH0252-5G
TCH0252-001G
TCH0252-100MG
5-Hydroxyindole ≥98.0%
5-Hydroxyindole
CAS Number: 1953-54-4
MDL Number: MFCD00005677
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 107
Formula:
C₈H₇NO MW: 133.15 g/mol Melting Pt: 107 °C |
MDL Number:
MFCD00005677 CAS Number: 1953-54-4 EINECS: 217-782-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...