Patton-Reeders reagent (Calconcarboxylic acid) (1:100)

Supplier: TCI America
Warning

Synonyms: 3-Hydroxy-4-(2-hydroxy-4-sulpho-1-naphthylazo)naphthalene-2-carboxylic acid, 3-Hydroxy-4-((2-hydroxy-4-sulfonaphthalen-1-yl)diazenyl)-2-naphthoic acid, Calconcarboxylic acid, 2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalene-3-carboxylic acid, 2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthoic acid, Cal-Red®, 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid, 3-Hydroxy-4-((2-hydroxy-4-sulphonaphthalen-1-yl)diazenyl)-2-naphthoic acid, 2-Hydroxy-1-(2-hydroxy-4-sulpho-1-naphthylazo)-3-naphthoic acid, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalene-2-carboxylic acid

H0250-25G H0250-1G
TCH0250-025GEA 130.05 CAD
TCH0250-025G TCH0250-001G
Patton-Reeders reagent (Calconcarboxylic acid) (1:100)
Patton-Reeders reagent (Calconcarboxylic acid)

CAS Number: 3737-95-9
MDL Number: MFCD00004078
Molecular Formula: C21H14N2O7S
Molecular Weight: 438.41
Form: Crystal
Color: Deep Blue


Other Product Names / Synonyms: Calcon-carboxylic Acid, Cal Red

Formula: C₂₁H₁₄N₂O₇S
MW: 438.42 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00004078
CAS Number: 3737-95-9
EINECS: 223-117-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0fe3689247db21cc8a5ce8e89421ae6d3b38417&vid=f8cf44dec59adc9ca98995955e0bad73a02ccf7e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR