1,2,4-Trihydroxybenzene ≥98.0%
Supplier: TCI America
H0249-25G
H0249-10G
TCH0249-25GEA
388.68
CAD
TCH0249-25G
TCH0249-010G
1,2,4-Trihydroxybenzene ≥98.0%
Formula:
C₆H₆O₃ Boiling Pt: 362 °C (1013 hPa) Melting Pt: 140 °C Density: 1.45 g/cm³ (20 °C) |
MDL Number:
MFCD00002198 CAS Number: 533-73-3 EINECS: 208-575-1 Merck Index: 13,01073 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...