4-Hexyloxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0247-25G H0247-500G
TCH0247-025GEA 120.08 CAD
TCH0247-025G TCH0247-500G
4-Hexyloxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Hexyloxybenzoic acid

CAS Number: 1142-39-8
MDL Number: MFCD00013991
Molecular Formula: C13H18O3
Molecular Weight: 222.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 155

Formula: C₁₃H₁₈O₃
Melting Pt: 155 °C
Density: ∼1 g/cm³ (20 °C)
MDL Number: MFCD00013991
CAS Number: 1142-39-8
EINECS: 214-534-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54543b7681f0f912e84da135baff40c5ef6e32c&vid=95d5c507690e4a6f52ea04d11e4c2bbc5eb8b566&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR