2-(2-Hydroxyethyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0246-500ML H0246-25ML
TCH0246-500MLEA 434.5 CAD
TCH0246-500ML TCH0246-025ML
2-(2-Hydroxyethyl)pyridine ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyethyl)pyridine

CAS Number: 103-74-2
MDL Number: MFCD00006364
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 92
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 2-(2-Hydroxyethyl)pyridine, 2-(2-Pyridyl)ethanol

Formula: C₇H₉NO
Boiling Pt: 139 °C
Melting Pt: –7.8 °C
Density: 1.10 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00006364
CAS Number: 103-74-2
EINECS: 203-140-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e6672ba99581ff5d7c688da22d71ec096020c8b&vid=6e6672ba99581ff5d7c688da22d71ec096020c8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR