N-(2-Hydroxyethyl)morpholine ≥99.0%

Supplier: TCI America

Synonyms: 4-(2-Hydroxyethyl)morpholine, 2-Morpholinoethanol

H0244-500ML H0244-25ML H0244-100ML
TCH0244-500MLEA 151.68 CAD
TCH0244-500ML TCH0244-025ML TCH0244-100ML
N-(2-Hydroxyethyl)morpholine ≥99.0%
N-(2-Hydroxyethyl)morpholine

CAS Number: 622-40-2
MDL Number: MFCD00006180
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Melting point (°C): 2
Flash Point (°C): 107
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Morpholinoethanol

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 222 °C
Melting Pt: 2 °C
Density: 1.08 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00006180
CAS Number: 622-40-2
EINECS: 210-734-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05b22a4aaa6f0fd9dacaa8df1720b39187c96378&vid=05b22a4aaa6f0fd9dacaa8df1720b39187c96378&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR