Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0241-25G H0241-500G
TCH0241-025GEA 33.18 CAD
TCH0241-025G TCH0241-500G
Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) ≥98.0% (by titrimetric analysis)
Isethionic acid sodium salt (Sodium 2-hydroxyethanesulphonate)

CAS Number: 1562-00-1
MDL Number: MFCD00007534
Molecular Formula: C2H6O4S
Molecular Weight: 148.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 194
Flash Point (°C): 100


Other Product Names / Synonyms: 2-Hydroxyethanesulfonic Acid Sodium Salt, Isethionic Acid Sodium Salt, Sodium 2-Hydroxyethanesulfonate

Formula: C₂H₅NaO₄S
Melting Pt: 194 °C
Flash Pt: 100 °C
MDL Number: MFCD00007534
CAS Number: 1562-00-1
EINECS: 216-343-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR