4-Benzylphenol ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
4-Hydroxydiphenylmethane, alpha-Phenyl-p-cresol
H0239-25G
H0239-500G
TCH0239-025GEA
129.56
CAD
TCH0239-025G
TCH0239-500G
4-Benzylphenol ≥98.0%
4-Benzylphenol
CAS Number: 101-53-1
MDL Number: MFCD00002384
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 330
Melting point (°C): 83
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Hydroxydiphenylmethane, alpha-Phenyl-p-cresol
Formula:
C₁₃H₁₂O MW: 184.24 g/mol Boiling Pt: 322 Melting Pt: 83 °C |
MDL Number:
MFCD00002384 CAS Number: 101-53-1 EINECS: 202-950-3 Merck Index: 13,01145 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...