7-Hydroxycoumarin ≥98.0%

Supplier: TCI America

H0236-25G H0236-5G
TCH0236-025GEA 91.64 CAD
TCH0236-025G TCH0236-005G
7-Hydroxycoumarin ≥98.0%
7-Hydroxycoumarin

CAS Number: 93-35-6
MDL Number: MFCD00006878
Molecular Formula: C9H6O3
Molecular Weight: 162.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 235


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 7-Hydroxycoumarin

Formula: C₉H₆O₃
Boiling Pt: 382 °C (1013 hPa)
Melting Pt: 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00006878
CAS Number: 93-35-6
EINECS: 202-240-3
Merck Index: 12,09981

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR