2-Hydroxychalcone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0234-5G H0234-25G
TCH0234-5GEA 96.38 CAD
TCH0234-5G TCH0234-025G
2-Hydroxychalcone ≥98.0% (by GC, titration analysis)
2-Hydroxychalcone

CAS Number: 644-78-0
MDL Number: MFCD00016449
Molecular Formula: C15H12O2
Molecular Weight: 224.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: 2-(2-Hydroxybenzal)acetophenone, 2-(2-Hydroxybenzylidene)acetophenone, Salicylalacetophenone

Formula: C₁₅H₁₂O₂
MDL Number: MFCD00016449
CAS Number: 644-78-0
EINECS: 211-422-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3167406db9893c032964c73c6f9ba2952c867f4&vid=d3167406db9893c032964c73c6f9ba2952c867f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR