4-Hydroxybenzyl alcohol ≥97.0%

Supplier: TCI America

H0224-25G H0224-100G
TCH0224-025GEA 74.78 CAD
TCH0224-025G TCH0224-100G
4-Hydroxybenzyl alcohol ≥97.0%
4-Hydroxybenzyl alcohol

CAS Number: 623-05-2
MDL Number: MFCD00004658
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Methylolphenol

Formula: C₇H₈O₂
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 115…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004658
CAS Number: 623-05-2
EINECS: 210-768-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bbf89cf74dc506d961fe7881f0c32588ee86644&vid=2bbf89cf74dc506d961fe7881f0c32588ee86644&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR