4-Hydroxybenzophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0222-500G H0222-25G
TCH0222-500GEA 496.12 CAD
TCH0222-500G TCH0222-025G
4-Hydroxybenzophenone ≥98.0% (by GC, titration analysis)
4-Hydroxybenzophenone

CAS Number: 1137-42-4
MDL Number: MFCD00002355
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134


Other Product Names / Synonyms: 4-Benzoylphenol

Formula: C₁₃H₁₀O₂
Boiling Pt: 367 °C (1013 hPa)
Melting Pt: 134 °C
Density: 1.133 g/cm³ (172 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002355
CAS Number: 1137-42-4
EINECS: 214-507-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=398412ebc56c5a5947bf9e4491ec3b88d9b7771c&vid=398412ebc56c5a5947bf9e4491ec3b88d9b7771c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR