Methyl-4-hydroxybenzoate 99,0

Supplier: TCI America
Warning

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

H0216-500G H0216-25G
TCH0216-500GEA 126.89 CAD
TCH0216-500G TCH0216-025G
Methyl-4-hydroxybenzoate 99,0
Methyl 4-hydroxybenzoate

CAS Number: 99-76-3
MDL Number: MFCD00002352
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 280
Melting point (°C): 127
Flash Point (°C): 96


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials, Bioscience, Antioxidants
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Methyl Ester, Methylparaben

Formula: HOC₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 280 °C
Melting Pt: 127 °C
Density: 1.1208 g/cm³ (137 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS Number: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22eedde41cc082d6247e560a68b6c6342ba8b124&vid=22eedde41cc082d6247e560a68b6c6342ba8b124&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR