Methyl-3-hydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester

H0215-500G H0215-25G H0215-100G
TCH0215-500GEA 810.54 CAD
TCH0215-500G TCH0215-025G TCH0215-100G
Methyl-3-hydroxybenzoate ≥98.0%
Methyl 3-hydroxybenzoate

CAS Number: 19438-10-9
MDL Number: MFCD00002295
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 73


Other Product Names / Synonyms: 3-Hydroxybenzoic Acid Methyl Ester

Formula: C₈H₈O₃
Boiling Pt: 178 °C
Melting Pt: 73 °C
MDL Number: MFCD00002295
CAS Number: 19438-10-9
EINECS: 243-071-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7d2a1418ec3519dec3c3c0b80963664ea75f08b&vid=d7d2a1418ec3519dec3c3c0b80963664ea75f08b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR