Heptyl-4-hydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: Heptylparaben, n-Heptyl 4-hydroxybenzoate, Heptyl parahydroxybenzoate, 4-Hydroxybenzoic acid heptyl ester

H0212-25G H0212-500G H0212-100G
TCH0212-025GEA 79 CAD
TCH0212-025G TCH0212-500G TCH0212-100G
Heptyl-4-hydroxybenzoate ≥98.0%
Heptyl 4-hydroxybenzoate

CAS Number: 1085-12-7
MDL Number: MFCD00016481
Molecular Formula: C14H20O3
Molecular Weight: 236.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 47


Other Product Names / Synonyms: Heptylparaben, 4-Hydroxybenzoic Acid Heptyl Ester

Formula: C₁₄H₂₀O₃
Melting Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00016481
CAS Number: 1085-12-7
EINECS: 214-115-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f98e2993ed292483ae5ff759f4627ad1fb165d4f&vid=f98e2993ed292483ae5ff759f4627ad1fb165d4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR