Ethyl 4-hydroxybenzoate ≥99.0% (by GC, titration analysis)

Supplier: TCI America

H0211-25G H0211-500G
TCH0211-025GEA 33.76 CAD
TCH0211-025G TCH0211-500G
Ethyl 4-hydroxybenzoate ≥99.0% (by GC, titration analysis)
Ethyl 4-hydroxybenzoate

CAS Number: 120-47-8
MDL Number: MFCD00002353
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 178
Melting point (°C): 117
Flash Point (°C): 107


Other Product Names / Synonyms: Ethylparaben, 4-Hydroxybenzoic Acid Ethyl Ester

Formula: C₉H₁₀O₃
Boiling Pt: 178 °C
Melting Pt: 117 °C
Density: 1.078 g/cm³ (131 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS Number: 120-47-8
EINECS: 204-399-4
Merck Index: 13,03869
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff9fa1b430130b67976c849d1e016d806c918cbf&vid=d92b68bc82745b396d5edc9bd40b831b3da16a34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR