Butyl-4-hydroxybenzoate ≥99.0% (by GC, titration analysis)
Supplier: TCI America
H0210-25G
H0210-500G
TCH0210-025GEA
48.06
CAD
TCH0210-025G
TCH0210-500G
Butyl-4-hydroxybenzoate ≥99.0% (by GC, titration analysis)
Butyl 4-hydroxybenzoate
CAS Number: 94-26-8
MDL Number: MFCD00016478
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 156
Melting point (°C): 70
Other Product Names / Synonyms: Butylparaben, 4-Hydroxybenzoic Acid Butyl Ester
Formula:
C₁₁H₁₄O₃ Boiling Pt: 156 °C Melting Pt: 70 °C Density: 1.23 g/cm³ (20 °C) |
MDL Number:
MFCD00016478 CAS Number: 94-26-8 EINECS: 202-318-7 Merck Index: 13,01583 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...