Pentyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0208-10G H0208-25G
TCH0208-010GEA 106.11 CAD
TCH0208-010G TCH0208-025G
Pentyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Pentyl 4-hydroxybenzoate

CAS Number: 6521-29-5
MDL Number: MFCD00016479
Molecular Formula: C12H16O3
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Freezing point (°C): 34
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Amyl Ester, Amylparaben, Pentyl 4-Hydroxybenzoate, 4-Hydroxybenzoic Acid Pentyl Ester

Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD00016479
CAS Number: 6521-29-5
EINECS: 229-408-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=169ebc8b5b1412aab1523d32d47453b732b136e7&vid=169ebc8b5b1412aab1523d32d47453b732b136e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR