4-Hydroxybenzoic acid 99,0

Supplier: TCI America
Warning

Synonyms: p-Hydroxybenzoic Acid

H0207-25G H0207-500G H0207-100G
TCH0207-025GEA 36.26 CAD
TCH0207-025G TCH0207-500G TCH0207-100G
4-Hydroxybenzoic acid 99,0
4-Hydroxybenzoic acid

CAS Number: 99-96-7
MDL Number: MFCD00002547
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 215

Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 336
Melting Pt: 215 °C
Density: 1.398 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002547
CAS Number: 99-96-7
EINECS: 202-804-9
Merck Index: 13,04837
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5ca2b03c5b8a6c340cd6589ee1050250f239dc3&vid=441d51cc7a4ef8e7af12673441593e37a8531ea7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR