Salicylic acid ≥99.5% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

H0206-25G H0206-500G
TCH0206-025GEA 46.06 CAD
TCH0206-025G TCH0206-500G
Salicylic acid ≥99.5% (by titrimetric analysis)
Salicylic acid

CAS Number: 69-72-7
MDL Number: MFCD00002439
Molecular Formula: C7H6O3
Molecular Weight: 138.12
Purity/Analysis Method: >99.5% (T)
Form: Crystal
Boiling point (°C): 211
Melting point (°C): 159
Flash Point (°C): 157


Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction), Bioscience, Other Plant Hormones, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: Salicylic Acid

Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 211 °C
Melting Pt: 159 °C
Density: 1.443 g/cm³ (20 °C)
Flash Pt: 157
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af19a13f90788031b22211fd9cd1a34c3d284cb0&vid=96dc472948b7b884b294697768e629a7bc9cf1c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR