4-Hydroxybenzamide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
H0200-25G
H0200-5G
TCH0200-025GEA
172.22
CAD
TCH0200-025G
TCH0200-005G
4-Hydroxybenzamide ≥98.0% (by total nitrogen basis)
4-Hydroxybenzamide
CAS Number: 619-57-8
MDL Number: MFCD00007997
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 161
Formula:
C₇H₇NO₂ Melting Pt: 161 °C |
MDL Number:
MFCD00007997 CAS Number: 619-57-8 EINECS: 210-602-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...