2'-Hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: o-Hydroxyacetophenone, 2-Acetophenol, 2-Hydroxyphenyl Methyl Ketone, 1-(2-Hydroxyphenyl)ethan-1-one, o-Acetylphenol, 1-(2-Hydroxyphenyl)-1-oxoethane, 1-Ethanoyl-2-hydroxybenzol, 2-Acetylphenol

H0192-25G H0192-500G H0192-100G
TCH0192-025GEA 53.72 CAD
TCH0192-025G TCH0192-500G TCH0192-100G
2'-Hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
2'-Hydroxyacetophenone

CAS Number: 118-93-4
MDL Number: MFCD00002219
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Boiling point (°C): 218
Melting point (°C): 6
Flash Point (°C): 110
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 2-Acetophenol, 2-Acetylphenol, 2-Hydroxyphenyl Methyl Ketone

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 218 °C
Melting Pt: 6 °C
Density: 1.13 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002219
CAS Number: 118-93-4
EINECS: 204-288-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83b9c4199ebed4a98afca098dacce47fb1c0452f&vid=83b9c4199ebed4a98afca098dacce47fb1c0452f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR