3-Phenylpropionic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3-Phenylpropanoic acid, Hydrocinnamic acid

H0183-25G H0183-500G H0183-100G
TCH0183-025GEA 40.2 CAD
TCH0183-025G TCH0183-500G TCH0183-100G
3-Phenylpropionic acid ≥98.0%
3-Phenylpropionic acid

CAS Number: 501-52-0
MDL Number: MFCD00002771
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 280
Melting point (°C): 48


Other Product Names / Synonyms: Hydrocinnamic Acid

Formula: C₉H₁₀O₂
Boiling Pt: 280 °C
Melting Pt: 48 °C
Density: 1.071 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
EINECS: 207-924-5
Merck Index: 13,04805
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93007619fd9bc8df4c82dfce56f2af9860c0e84e&vid=93007619fd9bc8df4c82dfce56f2af9860c0e84e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR