1-Cetylpyridinium bromide, hydrate ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H0162-25G H0162-500G
TCH0162-025GEA 43.42 CAD
TCH0162-025G TCH0162-500G
1-Cetylpyridinium bromide, hydrate ≥96.0% (by HPLC, titration analysis)
1-Cetylpyridinium bromide

CAS Number: 140-72-7 MDL Number: MFCD00011988 Molecular Formula: C21H38BrN Molecular Weight: 384.45 Purity/Analysis Method: 96.0% (HPLC,T) Form: Crystal


Product Categories: Materials Chemistry, Cationic Surfactants Other Product Names / Synonyms: Cetylpyridinium Bromide

Formula: C₂₁H₃₈BrN
MDL Number: MFCD00011988
CAS Number: 140-72-7
EINECS: 205-428-3
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=550fb2b4bcd0d9b069e89d945ebd0368f4122caf&vid=24b47e802e3e29f87b7b73aa2ad6191fe3b0d1ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR