Sodium hippurate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Hippuric Acid Sodium Salt, Sodium 2-benzamidoacetate

H0144-25G H0144-500G
TCH0144-025GEA 63.14 CAD
TCH0144-025G TCH0144-500G
Sodium hippurate ≥98.0% (by titrimetric analysis)
Sodium hippurate

CAS Number: 532-94-5
MDL Number: MFCD00002693
Molecular Formula: C9H9NO3
Molecular Weight: 201.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Benzoylglycine Sodium Salt, Hippuric Acid Sodium Salt

Formula: C₉H₈NNaO₃
MDL Number: MFCD00002693
CAS Number: 532-94-5
EINECS: 208-548-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83b5b64d594f4f8394a7c6c13592de0d63c5d80d&vid=70c7d6f21ab7d7da63dfc8ca3e434a7fe331ea21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR