Hippuric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

H0143-500G H0143-25G H0143-100G
TCH0143-500GEA 183.19 CAD
TCH0143-500G TCH0143-025G TCH0143-100G
Hippuric acid ≥98.0% (by titrimetric analysis)
Hippuric acid

CAS Number: 495-69-2
MDL Number: MFCD00002692
Molecular Formula: C9H9NO3
Molecular Weight: 179.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 190


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Benzoylglycine

Formula: C₆H₅C(O)NHCH₂COOH
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 190 °C
Density: 1.308 g/cm³ (20 °C)
MDL Number: MFCD00002692
CAS Number: 495-69-2
EINECS: 207-806-3
Merck Index: 13,04735
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cb6f570dc76232c8ba9dbbd8d49512ce4471867&vid=0cb6f570dc76232c8ba9dbbd8d49512ce4471867&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR