cis-3-Hexen-1-ol ≥97.0%
Supplier: TCI America
H0124-25ML
H0124-100ML
TCH0124-025MLEA
85.15
CAD
TCH0124-025ML
TCH0124-100ML
cis-3-Hexen-1-ol ≥97.0%
cis-3-Hexen-1-ol
CAS Number: 928-96-1
MDL Number: MFCD00063217
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 62
Specific Gravity (20/20): 0.85
Formula:
C₆H₁₂O Boiling Pt: 157 °C Melting Pt: –60 °C Density: 0.85 (20 °C) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063217 CAS Number: 928-96-1 EINECS: 213-192-8 UN: 1987 ADR: 3,III Merck Index: 13,04719 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...