Hexaphenyldisilane ≥97.0%

Supplier: TCI America
Warning

H0119-5G H0119-1G
TCH0119-5GEA 407.64 CAD
TCH0119-5G TCH0119-001G
Hexaphenyldisilane ≥97.0%
Hexaphenyldisilane

CAS Number: 1450-23-3
MDL Number: MFCD00039558
Molecular Formula: C36H30Si2
Molecular Weight: 518.81
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Flash Point (°C): 100

Formula: C₃₆H₃₀Si₂
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00039558
CAS Number: 1450-23-3
EINECS: 215-913-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc189e8a4a788ffb24b95256eaad021b9e031e2&vid=efc189e8a4a788ffb24b95256eaad021b9e031e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR