1,2,6-Hexanetriol ≥96.0%
Supplier: TCI America
H0104-25G
H0104-500G
TCH0104-025GEA
77.82
CAD
TCH0104-025G
TCH0104-500G
1,2,6-Hexanetriol ≥96.0%
1,2,6-Hexanetriol
CAS Number: 106-69-4
MDL Number: MFCD00002976
Molecular Formula: C6H14O3
Molecular Weight: 134.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 191
Specific Gravity (20/20): 1.10
Product Categories: Bioscience, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 1,2,6-Trihydroxyhexane
Formula:
C₆H₁₄O₃ Boiling Pt: 178 °C Melting Pt: 25…32 °C Density: 1.10 (20 °C) Flash Pt: 191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002976 CAS Number: 106-69-4 EINECS: 203-424-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...