Acetonylacetone ≥97.0%

Supplier: TCI America
Danger

Synonyms: Hexane-2,5-dione, 2,5-Hexanedione

H0101-25ML H0101-500ML H0101-100ML
TCH0101-025MLEA 71.1 CAD
TCH0101-025ML TCH0101-500ML TCH0101-100ML
Acetonylacetone ≥97.0%
Acetonylacetone

CAS Number: 110-13-4
MDL Number: MFCD00008792
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Melting point (°C): -9
Flash Point (°C): 79
Specific Gravity (20/20): 0.98


Other Product Names / Synonyms: 1,2-Diacetylethane, 2,5-Hexanedione

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 188 °C
Melting Pt: –9 °C
Density: 0.98 (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00008792
CAS Number: 110-13-4
EINECS: 203-738-3
Merck Index: 13,00072
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15e2ba330e3dd0559a83eb95f949c1b8195db43f&vid=b03a8441d554643c791acbe015bf5ea95faef13b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR