1,6-Hexanediol ≥97.0%

Supplier: TCI America

H0099-25G H0099-500G
TCH0099-025GEA 56.5 CAD
TCH0099-025G TCH0099-500G
1,6-Hexanediol ≥97.0%
1,6-Hexanediol

CAS Number: 629-11-8
MDL Number: MFCD00002985
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 208
Melting point (°C): 42
Flash Point (°C): 130


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,6-Dihydroxyhexane, Hexamethylene Glycol, 1,6-Hexylene Glycol

Formula: C₆H₁₄O₂
Boiling Pt: 208 °C
Melting Pt: 42 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00002985
CAS Number: 629-11-8
EINECS: 211-074-0
Merck Index: 13,04709
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313de57858f6aaf22c7dff14d5ab6875c7995330&vid=976c32f1e616e319797dbbe177b0c57498a01c18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR